missing translation for 'onlineSavingsMsg'
Learn More

Propamocarb 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30322353
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30322353 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30322353 Supplier Dr. Ehrenstorfer Supplier No. DREV16390000AL100
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Propamocarb free base
CAS 24579-73-5
Analyte or Component Names Propamocarb free base
Shipping Condition +20°C
Molecular Formula C9 H20 N2 O2
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Propamocarb free base
InChI Formula InChI=1S/C9H20N2O2/c1-4-8-13-9(12)10-6-5-7-11(2)3/h4-8H2,1-3H3,(H,10,12)
SMILES CCCOC(=O)NCCCN(C)C
IUPAC Name propyl N-[3-(dimethylamino)propyl]carbamate
Solvent or Matrix Acetonitrile
Industry Type Food Testing
Molecular Weight (g/mol) 188.27
Quantity 5 mL
Formula Weight 188.1525 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.