missing translation for 'onlineSavingsMsg'
Learn More

Propargyl bromide, 80 wt.% solution in toluene, stabilized

Product Code. 10665182 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
AcroSeal™ Glass bottle
Glass bottle
Unit Size:
1 L
250mL
50mL
This item is not returnable. View return policy

Product Code. 10665182

Brand: Thermo Scientific Acros 131482500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 106-96-7 | C3H3Br | 118.961 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material Propargyl bromide
Color White
CAS 106-96-7, 108-88-3
Boiling Point 88.0°C to 90.0°C
Molecular Formula C3H3Br
Linear Formula HC≡CCH2Br
Fieser 06,493; 10,332; 06,495
Density 1.3800g/mL
Synonym propargyl bromide, 3-bromopropyne, 3-bromo-1-propyne, 1-propyne, 3-bromo, 2-propynyl bromide, propynyl bromide, 1-bromo-2-propyne, propyne, 3-bromo, gamma-bromoallylene, 1-brom-2-propin
Solubility Information Solubility in water: immiscible.
InChI Key YORCIIVHUBAYBQ-UHFFFAOYSA-N
SMILES C#CCBr
IUPAC Name 3-bromoprop-1-yne
Molecular Weight (g/mol) 118.961
PubChem CID 7842
Formula Weight 118.96
Percent Purity 73 to 87% (propargyl bromide) (GC) 13 to 27% (toluene) (GC)
Packaging Glass bottle
Refractive Index 1.4900 to 1.4960
Specific Gravity 1.38
Quantity 250 mL
Flash Point 4°C
Physical Form Powder
Mostrar más Mostrar menos
compliance-icons compliance-icons compliance-icons compliance-icons
Nombre del producto
  • Propargyl bromide, 80 wt% solution in toluene, stabilized
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 3
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H361-Suspected of damaging fertility or the unborn child.
  • H373-May cause damage to organs.
  • H331-Toxic if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado