missing translation for 'onlineSavingsMsg'
Learn More

Propargyl bromide, 80 wt.% solution in toluene, stabilized, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 131480010

Code : 29

Additional Details : CAS Number : 106-96-7 Weight : 1.38000kg Transport : UN number : 3286 Chem class : 3 Pack group : II


 View more versions of this product

Product Code. 11944491

  • 410.00 € / 1 L
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
Specifications

Specifications

Propargyl bromide
106-96-7,108-88-3
C3H3Br
06,493; 10,332; 06,495
propargyl bromide, 3-bromopropyne, 3-bromo-1-propyne, 1-propyne, 3-bromo, 2-propynyl bromide, propynyl bromide, 1-bromo-2-propyne, propyne, 3-bromo, gamma-bromoallylene, 1-brom-2-propin
YORCIIVHUBAYBQ-UHFFFAOYSA-N
3-bromoprop-1-yne
7842
73 to 87% (propargyl bromide) (GC) 13 to 27% (toluene) (GC)
1.4900 to 1.4960
1 L
Powder
White
88.0°C to 90.0°C
HC≡CCH2Br
1.3800g/mL
Solubility in water: immiscible.
C#CCBr
118.961
118.96
AcroSeal™ Glass bottle
1.38
4°C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Propargyl bromide, 80 wt% solution in toluene, stabilized

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H361-Suspected of damaging fertility or the unborn child.
  • H373-May cause damage to organs.
  • H331-Toxic if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.

SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only