missing translation for 'onlineSavingsMsg'
Learn More

Propargyl chloride, 70% w/w soln. in toluene, Thermo Scientific™

Product Code. 15409169
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15409169 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15409169 Supplier Thermo Scientific Alfa Aesar Supplier No. L20416.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 624-65-7
Boiling Point 60°C to 63°C
Melting Point <-50°C
Molecular Formula C3H3Cl
Linear Formula HC≡CCH2Cl
MDL Number MFCD00000980
UN Number UN2924
Beilstein 506005
Concentration 70% w/w soln. in toluene
Density 0.99
Merck Index 14,7810
Synonym propargyl chloride, 3-chloropropyne, 3-chloro-1-propyne, 1-propyne, 3-chloro, 2-propynyl chloride, propyne, 3-chloro, propargylchloride, unii-3m62yfl252, gamma-chloroallylene, 1-chloro-2-propyne
Solubility Information Not miscible or difficult to mix with water.
InChI Key LJZPPWWHKPGCHS-UHFFFAOYSA-N
SMILES ClCC#C
IUPAC Name 3-chloroprop-1-yne
Molecular Weight (g/mol) 74.51
PubChem CID 12221
Formula Weight 74.51
Refractive Index 1.456
Quantity 100 g
Flash Point −15°C (−5°F)
Chemical Name or Material Propargyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.