missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Propidium Iodide

Product Code. 16488758
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488758 10 mg 10mg
16464677 50 mg 50mg
16438906 100 mg 100mg
16412326 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16488758 Supplier MedChemExpress Supplier No. HYD0815
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Propidium Iodide is a red-fluorescent dye that can be used to stain cells.

Specifications

Content And Storage 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Product Type Dye Reagents
Quantity 10 mg
Shipping Condition Room temperature
Solubility DMSO : 100 mg/mL (149.61 mM; Need ultrasonic) ∣ H2O : 3.57 mg/mL (5.34 mM; ultrasonic and warming and heat to 60°C)
Target HBV
Molecular Weight (g/mol) 668.39 g/mol
CAS 25535-16-4
Synonym PI
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.