missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Propionanilide, 98%, Thermo Scientific™

Product Code. 15434477
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15434477 5 g 5g
15444477 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15434477 Supplier Alfa Aesar™ Supplier No. H31832.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
The propionanilide derivatives are the first developed non-steroidal antiandrogens and include drugs such as flutamide, hydroxyflutamide, nilutamide and bicalutamide.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-71-3
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.193
MDL Number MFCD00092530
InChI Key ZTHRQJQJODGZHV-UHFFFAOYSA-N
Synonym propionanilide, n-phenylpropionamide, propanamide, n-phenyl, propionamide, n-phenyl, unii-uyp5zqi00t, uyp5zqi00t, methylacetanilid, n-phenyl-propionamide, ch3ch2conhph
PubChem CID 12107
IUPAC Name N-phenylpropanamide
SMILES CCC(=O)NC1=CC=CC=C1

Specifications

Melting Point 104°C to 107°C
Density 1.175
Boiling Point 222°C
Odor Odorless
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 149.19
Percent Purity 98%
Chemical Name or Material Propionanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.