missing translation for 'onlineSavingsMsg'
Learn More

Propylbenzene, 98%

Product Code. 10035680 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10035680 25 g Glass bottle 25g
10677211 100 g Glass bottle 100g
10558853 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10035680 Supplier Thermo Scientific Acros Supplier No. 418430250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103-65-1 | C9H12 | 120.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-65-1
Molecular Formula C9H12
Molecular Weight (g/mol) 120.20
MDL Number MFCD00009377
InChI Key ODLMAHJVESYWTB-UHFFFAOYSA-N
Synonym n-propylbenzene, 1-phenylpropane, phenylpropane, benzene, propyl, 1-propylbenzene, isocumene, n-propyl benzene, propyl-benzene, unii-0wr86zhg2z, propylbenzene, n
PubChem CID 7668
ChEBI CHEBI:42630
IUPAC Name propylbenzene
SMILES CCCC1=CC=CC=C1

Specifications

Melting Point -99°C
Color Yellow
Density 0.8600g/mL
Boiling Point 158°C
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2CH2CH3
Refractive Index 1.4900 to 1.493
Quantity 25 g
Beilstein 05, 390
Merck Index 15, 7957
Specific Gravity 0.86
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: miscible with ethanol,ether and acetone
Formula Weight 120.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Propylbenzene
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Propylbenzene
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.