missing translation for 'onlineSavingsMsg'
Learn More

Propylene Glycol 2-Beta-Glucopyranosiduronic Acid Benzyl Ester 2,3,4-Triacetate Ammonium Salt, TRC

Product Code. 30409883
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30409883 10 mg 10mg
30416698 25 mg 25mg
30421834 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30409883 Supplier Toronto Research Chemicals Supplier No. TRCP83522310MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Propylene Glycol 2-Beta-Glucopyranosiduronic Acid Benzyl Ester 2,3,4-Triacetate Ammonium Salt
Analyte or Component Names Propylene Glycol 2-Beta-Glucopyranosiduronic Acid Benzyl Ester 2,3,4-Triacetate Ammonium Salt
Shipping Condition Room Temperature
Molecular Formula C22H31NO11
InChI Formula InChI=1S/C22H28O11.H3N/c1-12(10-28-11-16-8-6-5-7-9-16)29-22-20(32-15(4)25)18(31-14(3)24)17(30-13(2)23)19(33-22)21(26)27;/h5-9,12,17-20,22H,10-11H2,1-4H3,(H,26,27);1H3/t12?,17-,18-,19-,20+,22+;/m0./s1
Solution Type Neat
SMILES CC(COCC1=CC=CC=C1)O[C@H]2[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@H](OC(C)=O)[C@@H](C([O-])=O)O2.[NH4+]
IUPAC Name ammonium (2S,3S,4S,5R,6R)-3,4,5-triacetoxy-6-((1-(benzyloxy)propan-2-yl)oxy)tetrahydro-2H-pyran-2-carboxylate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 485.48
Quantity 10 mg
Formula Weight 485.19
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.