missing translation for 'onlineSavingsMsg'
Learn More

Propylparaben, MedChemExpress

Product Code. 16482579
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482579 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16482579 Supplier Medchem Express Supplier No. HYN2026
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Propylparaben (Propyl parahydroxybenzoate) is an antimicrobial preservative which can be produced naturally by plants and bacteria. Propylparaben is prevalently used in cosmetics, pharmaceuticals, and foods. Propylparaben disrupts antral follicle growth and steroidogenic function by altering the cell-cycle, apoptosis, and steroidogenesis pathways. Propylparaben also decreases sperm number and motile activity in rats.

Propylparaben (Propyl parahydroxybenzoate) is an antimicrobial preservative which can be produced naturally by plants and bacteria. Propylparaben is prevalently used in cosmetics, pharmaceuticals, and foods. Propylparaben disrupts antral follicle growth and steroidogenic function by altering the cell-cycle, apoptosis, and steroidogenesis pathways. Propylparaben also decreases sperm number and motile activity in rats.

Specifications

Chemical Name or Material Propylparaben
Physical Form Solid
CAS 94-13-3
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C10H12O3
For Use With (Application) COVID-19-immunoregulation
Synonym Propyl parahydroxybenzoate Propyl 4-hydroxybenzoate
Solubility Information DMSO : 125 mg/mL (693.67 mM; Need ultrasonic)
SMILES O=C(OCCC)C1=CC=C(O)C=C1
Molecular Weight (g/mol) 180.2
Formula Weight 180.2
Percent Purity 98.93%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.