missing translation for 'onlineSavingsMsg'
Learn More

Propyphenazone, MedChemExpress

Product Code. 16419875
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16419875 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16419875 Supplier Medchem Express Supplier No. HYA0273
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Propyphenazone is a pyrazolone derivative with anti-inflammatory, analgesic and antipyretic activity, Propyphenazone-based analogues as prodrugs and selective cyclooxygenase-2 inhibitors.

Propyphenazone is a pyrazolone derivative with anti-inflammatory, analgesic and antipyretic activity, Propyphenazone-based analogues as prodrugs and selective cyclooxygenase-2 inhibitors.

Specifications

Chemical Name or Material Propyphenazone
Physical Form Solid
CAS 479-92-5
Color White
Quantity 250 mg
Purity Grade Notes Research
Molecular Formula C14H18N2O
For Use With (Application) Neuroscience-Neuromodulation
Synonym 4-Isopropylantipyrine Isopropylphenazone
Solubility Information DMSO : ≥ 2.6 mg/mL (11.29 mM)
SMILES O=C1N(C2=CC=CC=C2)N(C)C(C)=C1C(C)C
Molecular Weight (g/mol) 230.31
Formula Weight 230.31
Percent Purity 99.94%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.