missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Prostate Tissue Slides (Adenocarcinoma Gleasons Score 9)- Paraffin

Product Code. 18767773 Shop All Bio Techne Products
Change view
Click to view available options
Species:
Human
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Species Unit Size
18767773 Human Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18767773 Supplier R&D Systems™ Supplier No. NBP230303
  • ONLINE EXCLUSIVE PRICE
    323.30 €
    274.05 € / Pack of 5
    Save 49.25 €
    15% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems.

These slides are paraffin coated to prevent sample oxidization, it is recommended that slides are first de-paraffinized by baking at 60 degrees C for 1 hour in a vertical orientation prior to performing antigen retrieval procedures.

TRUSTED_SUSTAINABILITY

Specifications

Cross Reactivity Human
Type Prostate Tissue Slides (Adenocarcinoma Gleasons Score 9)
Regulatory Status RUO
Species Human
Tissue Condition Adenocarcinoma Gleasons Score 9
Product Line Novus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.