missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Prostatic Acid Phosphatase/ACPP Antibody Pair [HRP]

Product Code. p-7183367 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18215306

Brand: Novus Biologicals™ NBP27960815PLATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse Prostatic Acid Phosphatase/ACPP.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 156.25 pg/mL
Assay Sensitivity 156.25-10000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse Prostatic Acid Phosphatase/ACPP.
Target Species Mouse
For Use With (Application) Sandwich ELISA
Gene Alias acid phosphatase, prostate, ACP3, ACP-3, EC 3.1.3.2, PAP, prostatic acid phosphatase, prostatic acid phosphotase
Gene ID (Entrez) 55
Gene Symbol ACP3
Host Species Rabbit
Kit Contents
  • Standard: Each vial contains 810 ng of recombinant Mouse Prostatic Acid Phosphatase/ACPP.
  • Capture Antibody: 1 mg/mL of rabbit anti-Mouse Prostatic Acid Phosphatase/ACPP monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Prostatic Acid Phosphatase/ACPP monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Cell Cycle and Replication
    Storage Requirements Storage is content dependent.
    Target Prostatic Acid Phosphatase/ACPP
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.4 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 10000 pg/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.