missing translation for 'onlineSavingsMsg'
Learn More

PROTAC ERα Degrader-1, MedChemExpress

Product Code. 16474568
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
Unit Size:
10mg
2mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16474568 10 mg 10mg
16417287 5 mg 5mg
16410146 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16474568 Supplier Medchem Express Supplier No. HY112098
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress PROTAC ERα Degrader-1 comprises an ubiquitin E3 ligase binding group, a linker and a protein binding group. PROTAC ERα Degrader-1 extracts from patent WO2017201449A1, compound P1. PROTAC ERα Degrader-1 is an estrogen receptor-alpha (ERα) degrader.

PROTAC ERα Degrader-1 comprises an ubiquitin E3 ligase binding group, a linker and a protein binding group. PROTAC ERα Degrader-1 extracts from patent WO2017201449A1, compound P1. PROTAC ERα Degrader-1 is an estrogen receptor-alpha (ERα) degrader.

Specifications

Chemical Name or Material PROTAC ERα Degrader-1
CAS 2417369-94-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C66H69N7O10
Quantity 10 mg
Solubility Information DMSO : 200 mg/mL (178.53 mM; Need ultrasonic)
SMILES OC1=CC=C(/C(C2=CC=C(OCCN(C)C(COCCOCCNC(C3=CC=C(C(N4[C@@H](C)[C@H](NC([C@H](C)NC)=O)C(N(CC5=C(OC)C=CC6=C5C=CC=C6)C7=C4C=C(C#N)C=C7)=O)=O)C=C3)=O)=O)C=C2)=C(C8=CC=CC=C8)\CC)C=C1
Molecular Weight (g/mol) 1120.29
Formula Weight 1120.29
Percent Purity 95.32%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.