missing translation for 'onlineSavingsMsg'
Learn More

PROTAC ERRα ligand 1, MedChemExpress

Product Code. 16449306
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16449306 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16449306 Supplier Medchem Express Supplier No. HYU00425
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress PROTAC ERRα ligand 1 is an estrogen-related receptor α (ERRα) antagonist with IC50s of 0.04 and 2.8 μM for ERRα and ERRγ, respectively.

PROTAC ERRα ligand 1 is an estrogen-related receptor α (ERRα) antagonist with IC50s of 0.04 and 2.8 μM for ERRα and ERRγ, respectively.

Specifications

Chemical Name or Material PROTAC ERRα ligand 1
CAS 1264754-13-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H11F3N2O4S
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (237.89 mM; Need ultrasonic)
SMILES N#CC1=CC=C(OC2=CC=C(/C=C(SC(N3)=O)/C3=O)C=C2OC)C(C(F)(F)F)=C1
Molecular Weight (g/mol) 420.36
Formula Weight 420.36
Percent Purity 98.12%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.