missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteasome 19S S3 Antibody (OTI2A2), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73636DL594
This item is not returnable.
View return policy
Description
Proteasome 19S S3 Monoclonal antibody specifically detects Proteasome 19S S3 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry
Specifications
| Proteasome 19S S3 | |
| Monoclonal | |
| DyLight 594 | |
| 26S proteasome regulatory subunit RPN3, 26S proteasome regulatory subunit S3, P58, proteasome (prosome, macropain) 26S subunit, non-ATPase, 3, Proteasome subunit p58, RPN3, S3, 26S proteasome non-ATPase regulatory subunit 3, tissue specific transplantation antigen 2, TSTA2 | |
| Full length recombinant protein of human Proteasome 19S S3 (NP_002800) produced in HEK293T cells. | |
| 0.1 mL | |
| Neuroscience, Ubiquitin Proteasome Pathway | |
| 5709 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Flow Cytometry | |
| OTI2A2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction