missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteasome beta 1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93490-0.02ml
This item is not returnable.
View return policy
Description
Proteasome beta 1 Polyclonal antibody specifically detects Proteasome beta 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Proteasome beta 1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| EC 3.4.25.1, FLJ25321, HC5, KIAA1838, Macropain subunit C5, Multicatalytic endopeptidase complex subunit C5, PMSB1, proteasome (prosome, macropain) subunit, beta type, 1, proteasome beta 1 subunit, Proteasome component C5, Proteasome gamma chain, proteasome subunit beta type-1, proteasome subunit HC5, PSC5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 29-241 of human PSMB1 (NP_002784.1). RFSPYVFNGGTILAIAGEDFAIVASDTRLSEGFSIHTRDSPKCYKLTDKTVIGCSGFHGDCLTLTKIIEARLKMYKHSNNKAMTTGAIAAMLSTILYSRRFFPYYVYNIIGGLDEEGKGAVYSFDPVGSYQRDSFKAGGSASAMLQPLLDNQVGFKNMQNVEHVPLSLDRAMRLVKDVFISAAERDVYTGDALRICIVTKEGIREETVSLRKD | |
| 0.02 mL | |
| Cell Biology | |
| 5689 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction