Learn More
Invitrogen™ Protein A Recombinant Rabbit Monoclonal Antibody (8L5X0)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557986
Description
Immunogen sequence: AQHDEAQQNA FYQVLNMPNL NADQRNGFIQ SLKDDPSQSA NVLGEAQKLN DSQAPKADAQ QNNFNKDQQS AFYEILNMPN LNEAQRNGFI QSLKDDPSQS TNVLGEAKKL NESQAPKADN NFNKEQQNAF YEILNMPNLN EEQRNGFIQS LKDDPSQSAN LLSEAKKLNE SQAPKADNKF NKEQQNAFYE ILHLPNLNEE QRNGFIQSLK DDPSQSANLL AEAKKLNDAQ APKADNKFNK EQQNAFYEIL HLPNLTEEQR NGFIQSLKDD PSVSKEILAE AKKLNDAQAP KEEDNNKPGK EDNNKPGKED NNKPGKEDNN KPGKEDNNKP GKEDGNKPGK EDNKKPGKED GNKPGKEDNK KPGKEDGNKP GKEDGNKPGK EDGNGVHVVK PGDTVNDIAK ANGTTADKIA ADNKLADKNM IKPGQELVVD KKQPANHADA NKAQALPET.
Protein A is an Orphan-U GPCR with an unknown ligand. This gene was identified from genomic analysis of a gene-dense region at human chromosome 12p13. It appears to be expressed mainly in the brain. However, its function is not known. Three alternatively spliced variants have been described; the full length of the third variant is yet to be determined.
Specifications
| Protein A | |
| Recombinant Monoclonal | |
| 1.11 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 37-485 of Staphylococcus aureus Protein A (YP_498670.1). | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 8L5X0 | |
| Unconjugated | |
| Rabbit | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.