Learn More
Invitrogen™ Protein APC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596213
Description
Immunogen sequence: QSKENQVSAK GTWRKIKENE FSPTNSTSQT VSSGATNGAE SKTLIYQMAP AVSKTEDVWV RIEDCPINNP RSGRSPTGNT PPVIDSVSEK ANPNIKDSKD NQAKQNVGNG SVPMRTVGLE NRLNSFIQVD APDQKGTEIK PGQNNPVPVS ETNESSIVER T Positive Samples: HeLa; Cellular Location: Cell junction, Cell membrane, Cell projection, Cytoplasm, adherens junction, cytoskeleton, lamellipodium, ruffle membrane.
This gene encodes a tumor suppressor protein that acts as an antagonist of the Wnt signaling pathway. It is also involved in other processes including cell migration and adhesion, transcriptional activation, and apoptosis. Defects in this gene cause familial adenomatous polyposis (FAP), an autosomal dominant pre-malignant disease that usually progresses to malignancy. Disease-associated mutations tend to be clustered in a small region designated the mutation cluster region (MCR) and result in a truncated protein product.
Specifications
| Protein APC | |
| Polyclonal | |
| Unconjugated | |
| Apc | |
| adenomatosis polyposis coli; adenomatosis polyposis coli tumor suppressor; adenomatous polyposis coli; adenomatous polyposis coli protein; AI047805; APC; APC, WNT signaling pathway regulator; AU020952; AW124434; BTPS2; CC1; Deleted in polyposis 2.5; DP2; DP2.5; DP3; FAP; FPC; GS; mAPC; Min; multiple intestinal neoplasia; PPP1R46; Protein APC; protein phosphatase 1, regulatory subunit 46; RATAPC; truncated adenomatosis polyposis coli; WNT signaling pathway regulator | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11789, 24205, 324 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| 1.99 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P25054, P70478, Q61315 | |
| Apc | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 2600-2760 of human APC (NP_0000292). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.