missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Protein Expression Medium (PEM)
GREENER_CHOICE

Product Code. 10621864
Change view
Click to view available options
Quantity:
1 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10621864 1 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10621864 Supplier Gibco™ Supplier No. 12661013

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Protein Expression Medium (PEM)

  • Supports the large-scale, high-density culture of transfected PER.C6™ cells in shake flasks, roller bottles, and bioreactors
  • Contains no components directly derived from human or animal sources and one low-level, recombinant protein, minimizing the opportunity to introduce adventitious agents
  • Formulated without L-glutamine for maximum stability
  • Supports parental PER.C6™, HEK 293, and HeLa cell growth in suspension culture, as well as transfected PER.C6™ cells for protein expression applications
  • Store in the dark at 2-8°C. PER.C6™ is a trademark of Crucell, Holland B.V.
TRUSTED_SUSTAINABILITY

Specifications

Cell Line HeLa, PER.C6, 293 (HEK)
Classification Animal Origin-free, Low Protein, Serum-free
Concentration 1 X
Form Liquid
Product Type Protein Expression Medium (PEM)
Serum Level Serum-free
With Additives Sodium Bicarbonate
Without Additives No Glutamine, No Phenol Red, No Sodium Pyruvate
Product Line Gibco
Quantity 1 L
Shipping Condition Room Temperature
Endotoxin Level Low
Format Bottle(s)
Included Antibiotics No antibiotics
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Culture Type Suspension Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.