missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Protein LoBind™ Deepwell Plates
BIOPHARMA

Product Code. 10574073 Shop All Eppendorf Products
Change view
Click to view available options
Color:
White
White, Clear Bottom
Yellow
No. of Wells:
96
Volume (Metric) Well:
500 μL
1000 μL
2000 μL
Packaging:
10 Bags x 12 Plates
10 Bags x 8 Plates
5 Bags x 4 Plates
5 Bags x 8 Plates
Unit Size:
Pack of 120
Pack of 20
Pack of 40
Pack of 80
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color No. of Wells Volume (Metric) Well Packaging Unit Size
10574073 Yellow 96 500 μL 5 Bags x 8 Plates Pack of 40
10367133 White 96 500 μL 10 Bags x 12 Plates Pack of 120
10709263 White 96 1000 μL 5 Bags x 4 Plates Pack of 20
10215373 White, Clear Bottom 96 1000 μL 10 Bags x 8 Plates Pack of 80
11518643 White 96 2000 μL 5 Bags x 4 Plates Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10574073 Supplier Eppendorf™ Supplier No. 0030504119
  • ONLINE EXCLUSIVE PRICE
    889.43 €
    789.00 € / Pack of 40
    Save 100.43 €
    11% Off
In Stock
Add to basket
Add to basket
Bulk Quote Request

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Eppendorf Protein LoBind Plates are designed for use in proteomics or other fields of protein research, where protein concentration tend to be very small and sample recovery is vital for assay results.

  • Eppendorf LoBind material ensures excellent sample recovery for improved assay results.
  • Free of surface coating (e.g., silicone) to minimize the risk of sample interference.
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors.
  • High-contrast Unique OptiTrack™ matrix: up to 30% faster sample identification and fewer pipetting errors.
  • RecoverMax™ well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties.
  • Raised well rims and a smooth surface ensure reliable sealing in plates.
TRUSTED_SUSTAINABILITY

Specifications

Certified Yes
Detection Method Quantitative Protein Analysis
Array 8 x 12
Autoclavable Autoclavable
Packaging 5 Bags x 8 Plates
Color Yellow
Product Type Protein LoBind™ Plate
Material Polypropylene
Volume (Metric) Well 500 μL
No. of Wells 96
Sterility Non-sterile
Dnase Free Yes
For Use With (Application) Preparation or Storage of Protein, Peptide or Antibody Samples
Max. Heat Stability 121°C
Max. RCF 6000 x g
Profile Deep well plates, 96 Wells
Purity or Quality Grade PCR Clean (Non-sterile)
Quantity 40 Pk.
Well Color Colorless
Dimensions (L x W x H) 85.5 x 127.8 x 27.1 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.