missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein O-Glucosyltransferase 1/POGLUT1/KTELC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP1-90311-25ul
This item is not returnable.
View return policy
Description
Protein O-Glucosyltransferase 1/POGLUT1/KTELC1 Polyclonal specifically detects Protein O-Glucosyltransferase 1/POGLUT1/KTELC1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Protein O-Glucosyltransferase 1/POGLUT1/KTELC1 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500-1:1000 | |
| 9630046K23Rik, C3orf9, CAP10-like protein, 46 kDa, chromosome 3 open reading frame 9, CLP46, EC 2.4.1.-, hCLP46CAP10-like 46 kDa protein, KDELC family like 1, KDELCL1, KTEL (Lys-Tyr-Glu-Leu) containing 1, KTEL motif-containing protein 1, MDS010, MDSRPKTELC1, MGC32995, Myelodysplastic syndromes relative protein, protein O-glucosyltransferase 1, Rumi, x 010 protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| POGLUT1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:VQELLQFVKANDDVAQEIAERGSQFIRNHLQMDDITCYWENLLSEYSKFLSYNVTRRKGYDQIIPKMLKTEL | |
| 25 μL | |
| Developmental Biology | |
| 56983 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction