missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Protein Phosphatase 1C gamma Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30769-25ug
This item is not returnable.
View return policy
Description
Protein Phosphatase 1C gamma Polyclonal antibody specifically detects Protein Phosphatase 1C gamma in Bovine,Human,Mouse,Rat samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| Protein Phosphatase 1C gamma | |
| Polyclonal | |
| Western Blot : 1:500-1:1000, Immunoprecipitation | |
| EC 3.1.3.16, PP-1G, PP1gamma, PPP1G, protein phosphatase 1, catalytic subunit, gamma isoform, protein phosphatase 1, catalytic subunit, gamma isozyme, Protein phosphatase 1C catalytic subunit, serine/threonine phosphatase 1 gamma, serine/threonine-protein phosphatase PP1-gamma catalytic subunit | |
| The immunogen used to generate the purified antibody was a peptide conjugated to KLH corresponding to the sequence NH2-Thr-Pro-Pro-Arg-Gly-Met-Ile-Thr-Lys-Gln-Ala-Lys-Lys-COOH. This peptide antibody corresponds to C -terminal peptide of Protein Phosphatase 1C gamma catalytic subunit having a MW of 37kD. The sequence used is amino acid 311-323. | |
| 25 μg | |
| Cancer, Lipid and Metabolism, Protein Phosphatase, Stem Cell Markers | |
| 5501 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Sheep | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Bovine, Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction