Learn More
Novus Biologicals™ Protein Phosphatase 1C gamma Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-30770-100ug
 View more versions of this product
Product Code. 30067095
  • 508.00 € / 100µg
Description
Protein Phosphatase 1C gamma Polyclonal antibody specifically detects Protein Phosphatase 1C gamma in Bovine,Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Protein Phosphatase 1C gamma
Polyclonal
Western Blot : 10 μg/mL
EC 3.1.3.16, PP-1G, PP1gamma, PPP1G, protein phosphatase 1, catalytic subunit, gamma isoform, protein phosphatase 1, catalytic subunit, gamma isozyme, Protein phosphatase 1C catalytic subunit, serine/threonine phosphatase 1 gamma, serine/threonine-protein phosphatase PP1-gamma catalytic subunit
QKASNYRNNTVKYE
100 μg
Cancer, Lipid and Metabolism, Protein Phosphatase, Stem Cell Markers
5501
Store at -20°C. Avoid freeze/thaw cycles.
IgG
Western Blot
Unconjugated
Phosphate buffered saline
Sheep
Ammonium sulfate precipitation
RUO
Primary
Bovine, Human, Mouse, Rat
Purified