missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein phosphatase 1F Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00 € - 572.00 €
Specifications
| Antigen | Protein phosphatase 1F |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18443801
|
Novus Biologicals
NBP1-88206-25ul |
25 μL |
280.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18212056
|
Novus Biologicals
NBP1-88206 |
0.1 mL |
572.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Protein phosphatase 1F Polyclonal specifically detects Protein phosphatase 1F in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Protein phosphatase 1F | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Protein Phosphatase | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 9647 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:VVPHQEVVGLVQSHLTRQQGSGLRVAEELVAAARERGSHDNITVMVVFLRDPQELLEGGNQGEGDPQAEGRRQDLPSSLPEPETQ | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| Ca(2+)/calmodulin-dependent protein kinase phosphatase, CaMKPaseCaM-kinase phosphatase, EC 3.1.3.16, FEM-2, hFEM-2, KIAA0015POPX2CAMKP, Partner of PIX 2, PP2C phosphatase, Protein fem-2 homolog, protein phosphatase 1F, protein phosphatase 1F (PP2C domain containing), protein phosphatase, Mg2+/Mn2+ dependent, 1F | |
| PPM1F | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title