missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein Phosphatase inhibitor 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24490
This item is not returnable.
View return policy
Description
Protein Phosphatase inhibitor 1 Polyclonal specifically detects Protein Phosphatase inhibitor 1 in Human, Canine, Equine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Protein Phosphatase inhibitor 1 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.2% Gelatin with 0.05% Sodium Azide | |
| I1, I-1, IPP-1, IPP1inhibitor-1, protein phosphatase 1 regulatory subunit 1A, protein phosphatase 1, regulatory (inhibitor) subunit 1A, Protein phosphatase inhibitor 1, protein phosphatase inhibitor-1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human, monkey, rat, mouse, cowand chimpanzee and 73% homologous in chicken. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 0.5-2 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml | |
| Q13522 | |
| PPP1R1A | |
| A portion of amino acids 50-100 of human IPP-1 was used as the immunogen. | |
| 0.1 mg | |
| Signal Transduction | |
| 5502 | |
| Human, Canine, Equine, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction