missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Proteome Purify 12 Human Serum Protein Immunodepletion Resin

Product Code. 18481280
Change view
Click to view available options
Quantity:
20 Tests
40 Tests
Unit Size:
20mL
40mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18481280 20 Tests 20mL
18472270 40 Tests 40mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18481280 Supplier R&D Systems™ Supplier No. IDR012020
  • ONLINE EXCLUSIVE PRICE
    662.00 €
    625.80 € / 20mL
    Save 36.20 €
    5% Off
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Immunodepletion Resin, Spin-X Filter Units

A single-use alternative to existing column or spin cartridge systems to deplete serum or plasma of high abundance proteins

Depletes samples of the 12 high-abundance proteins comprising approximately 95% of proteins in human serum or plasma
Depletes samples of albumin, IgA, IgG and IgM, Fibrinogen, Transferrin, Haptoglobin and others
Removal allows visualization of proteins

TRUSTED_SUSTAINABILITY

Specifications

Quantity 20 Tests
Product Type Immunodepletion Resin
Content And Storage Store the unopened product at 2 - 8° C. Do not use past expiration date.
Product Line R&D Systems
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.