missing translation for 'onlineSavingsMsg'
Learn More

Prothoate, Dr. Ehrenstorfer

Product Code. 30316215
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316215 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316215 Supplier Dr. Ehrenstorfer Supplier No. DREC16570000
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Prothoate
CAS 2275-18-5
Analyte or Component Names Prothoate
Shipping Condition Room Temperature
Molecular Formula C9 H20 N O3 P S2
Certifications/Compliance ISO 17034
Synonym Phosphorodithioic acid, O,O-diethyl ester, S-ester with N-isopropyl-2-mercaptoacetamide (6CI,7CI,8CI), Acetamide, N-isopropyl-2-mercapto-, S-ester with O,O-di-Et phosphorodithioate (6CI), American Cyanamid 18,682, FAC, FAC (pesticide), Fac 20, Fak 40, Fostion, O,O-Diethyl S-(N-isopropylcarbamoylmethyl) phosphorodithioate, Prothoat, Prothoate, Trimethoate
InChI Formula InChI=1S/C9H20NO3PS2/c1-5-12-14(15,13-6-2)16-7-9(11)10-8(3)4/h8H,5-7H2,1-4H3,(H,10,11)
Solution Type Neat
SMILES CCOP(=S)(OCC)SCC(=O)NC(C)C
IUPAC Name 2-diethoxyphosphinothioylsulfanyl-N-propan-2-ylacetamide
Industry Type Environmental Testing
Molecular Weight (g/mol) 285.36
Quantity 25 mg
Formula Weight 285.0622 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.