missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prothrombin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
214.00 € - 584.00 €
Specifications
| Antigen | Prothrombin |
|---|---|
| Dilution | Western Blot 1:100 - 1:500, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30228623
|
Novus Biologicals
NBP3-35241-20ul |
20 μL |
214.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30231452
|
Novus Biologicals
NBP3-35241-100ul |
100 μL |
584.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Prothrombin Polyclonal antibody specifically detects Prothrombin in Human samples. It is validated for ELISA,Western BlotSpecifications
| Prothrombin | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Breast Cancer, Cancer | |
| PBS (pH 7.3), 50% glycerol | |
| 2147 | |
| IgG | |
| Affinity purified |
| Western Blot 1:100 - 1:500, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Coagulation factor II, coagulation factor II (thrombin), EC 3.4.21, EC 3.4.21.5, prothrombin, prothrombin B-chain, PT, serine protease | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 364-622 of human Prothrombin (NP_000497.1).,, Sequence:, IVEGSDAEIGMSPWQVMLFRKSPQELLCGASLISDRWVLTAAHCLLYPPWDKNFTENDLLVRIGKHSRTRYERNIEKISMLEKIYIHPRYNWRENLDRDIALMKLKKPVAFSDYIHPVCLPDRETAASLLQAGYKGRVTGWGNLKETWTANVGKGQPSVLQVVNLPIVERPVCKDSTRIRITDNMFCAGYKPDEGKRGDACEGDSGGPFVMKSPFNNRWYQMGIVSWGEGCDRDGKYGFYTHVFRLKKWIQKVIDQFGE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title