missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prox1 Antibody (CL13474) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24561JF669
This item is not returnable.
View return policy
Description
Prox1 Monoclonal antibody specifically detects Prox1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| Prox1 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| Homeobox prospero-like protein PROX1, prospero homeobox 1, prospero homeobox protein 1, prospero-related homeobox 1, PROX-1 | |
| This antibody was developed using a recombinant protein derived from Q92786, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Vision | |
| 5629 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunocytochemistry/Immunofluorescence | |
| CL13474 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction