missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hamilton™ PRP-1 7 μm 100 Å 21.2 x 250 mm
Click to view available options
Length (Metric):
250 mm
Unit Size:
Each
Description
Specification Sheet
- Separation Mode – Reversed-Phase
- Stationary Phase – PRP-1
- Format – HPLC Column
- Hardware Inner Diameter – 21.2 mm
- Hardware Length – 250 mm
- Particle Size – 7 μm
- USP Classification – USP L21
- Packing Material Type – PSDVB
- Pore Size – 100 Å
- Hardware Material – Stainless Steel
- Max Pressure – 5,000 psi
- Mobile Phase Limits – pH 1-13. 0-100% aqueous, organic modifier
- Buffer Strength – 0.0-0.5N
- Temp Limits – 5-85°C
- Restoration – Run a gradient of 100% water to 100% acetonitrile. Repeat three times
- Compound Classification – Organics
- UOM – 1 EACH
Specifications
Specifications
| Column Format | HPLC Column |
| Diameter (Metric) | 21.2 mm |
| Length (Metric) | 250 mm |
| Max. Pressure | 5,000 psi |
| Particle Size | 7 μm |
| pH | pH 1-13. 0-100% aqueous, organic modifier |
| Pore Size | 100 Å |
| Temperature | 5° to 85°C |
| USP Type | USP L21 |
| Packing Material | PSDVB |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction