missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PrPC Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
310.00 € - 500.00 €
Specifications
| Antigen | Prion protein |
|---|---|
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18670786
|
Novus Biologicals
NBP2-38508-25ul |
25 μL |
310.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18178588
|
Novus Biologicals
NBP2-38508 |
0.1 mL |
500.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PrPC Polyclonal specifically detects PrPC in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Prion protein | |
| Polyclonal | |
| Rabbit | |
| Neuroscience, Stem Cell Markers | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| CD230, CJD, fatal familial insomnia), GSS, prion protein, prion protein (p27-30), prion protein PrP, prion-related protein, PRIPMGC26679 | |
| PRNP | |
| IgG | |
| Affinity Purified |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| P04156 | |
| 5621 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: GSPGGNRYPPQGGGGWGQPHGGGWGQPHGGGWGQPHGGGWGQPHGGGWGQGGGTHSQWNKPSKP | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title