missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRPF4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12635-50ul
This item is not returnable.
View return policy
Description
PRPF4 Polyclonal antibody specifically detects PRPF4 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| PRPF4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-5ul/mg of lysate | |
| hPrp4, HPRP4P, HPRP4PRP4/STK/WD splicing factor, PRP4 homolog, PRP4 pre-mRNA processing factor 4 homolog (yeast), Prp4p, PRP4WD splicing factor Prp4, U4/U6 small nuclear ribonucleoprotein Prp4, U4/U6 snRNP 60 kDa protein | |
| Produced in rabbits immunized with E. coli-derived Human PRPF4 fragment | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 9128 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction