Learn More
Invitrogen™ PRPF4 Recombinant Rabbit Monoclonal Antibody (24GB975)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557618
 View more versions of this product
Product Code. 30276235
  • 564.00 € / 100µL
Description
PRPF4 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The removal of introns from nuclear pre-mRNAs occurs on complexes called spliceosomes, which are made up of 4 small nuclear ribonucleoprotein (snRNP) particles and an undefined number of transiently associated splicing factors. PRPF4 is 1 of several proteins that associate with U4 and U6 snRNPs.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PRPF4
Recombinant Monoclonal
Unconjugated
PRPF4
1600015H11Rik; AI874830; AW047464; bN189G18.1; HPRP4; HPRP4P; hypothetical protein LOC508735; mg:ab03a02; pre-mRNA processing factor 4; PRP4; PRP4 homolog; PRP4 pre-mRNA processing factor 4 homolog; PRP4 pre-mRNA processing factor 4 homolog (yeast); PRP4/STK/WD splicing factor; Prp4p; PRPF4; RP70; SNRNP60; U4/U6 small nuclear ribonucleoprotein Prp4; U4/U6 snRNP 60 kDa protein; WD splicing factor Prp4; zgc:65943
Rabbit
Affinity chromatography
RUO
298095, 70052, 9128
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB975
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O43172, Q9DAW6
PRPF4
A synthesized peptide derived from human PRPF4 (aa 1-50).
100 μL
Primary
Human, Mouse, Rat