missing translation for 'onlineSavingsMsg'
Learn More

pS2/pNR-2 Estrogen-Regulated Protein Ab-1, Mouse Monoclonal Antibody,Lab Vision™ Epredia™

Provide accurate, reproducible results with the Epredia™ pS2/pNR-2 Estrogen-Regulated Protein Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia MS-111-P1ABX

 View more versions of this product

Product Code. 12689026

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

pS2 is a cysteine-rich, 6.5kDa protein found in both estrogen-dependent (breast tumors) and estrogen-independent tissues (normal stomach mucosa). About 60% of breast carcinomas are positive for pS2. Staining is cytoplasmic, often with localization to the Golgi apparatus. pS2 is primarily expressed in estrogen receptor-positive breast tumors.

Host Species: Mouse

Clone: pS2.1

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: aa 54-84

Immunogen: A synthetic 31-mer peptide aa 54-84 (KGCCFDDTVRGVPWCFYPNTIDVPPEEECEF) from the C-terminus of human pS2 protein

Molecular Weight: 6.5kDa

Cellular Localization: Cytoplasmic Antibody to pS2 is reportedly useful in identifying a subset of estrogen-dependent breast tumors which may respond to endocrine therapy.

Recommended for:

  • Immunohistochemistry

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

pS2/pNR-2 Estrogen-Regulated Protein Ab-1
Monoclonal
1mg/mL
Mouse
100 μL
Cancer and Tumor Biology
Human
IgG1
Immunohistochemistry (Paraffin)
pS2.1
Unconjugated
A Synthetic 31-mer peptide aa 54-84 (KGCCFDDTVRGVPWCFYPNTIDVPPEEECEF) from the C-terminus of human pS2 protein
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.