Learn More
Invitrogen™ PSA Monoclonal Antibody (A67-B/E3), Biotin
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA110447
Product Code. 12362443
  • 378.00 € / 100µg
Description
This antibody recognizes both free and complexed PSA.
PSA is a chymotrypsin-like serine protease (kallikrein family) produced by the prostate epithelium, and is abundant in seminal fluid. PSA can be detected in the sera of patients with prostatic carcinoma. It is predominantly complexed to a liver-derived serine protease inhibitor, alpha-1-antichymotrypsin (ACT). A higher proportion of serum PSA is complexed to ACT in prostate cancer than in benign prostate hyperplasia. PSA is used to confirm prostatic acinar cell origin in primary and metastatic carcinoma and to rule out non-prostatic carcinoma mimics.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PSA
Monoclonal
1 mg/mL
PBS with 15mM sodium azide; pH 7.4
P07288
KLK3
30 KDa protein fraction prepared from human seminal plasma.
100 μg
Primary
Human
Antibody
IgG1
ELISA, Immunohistochemistry (Paraffin), Western Blot
A67-B/E3
Biotin
KLK3
Antigen; antigen, prostate specific; APS; Fletcher factor; gamma-seminoprotein; hK3; Kal3; Kal-3; Kallikrein 3; kallikrein 3, plasma; kallikrein B, plasma 1; kallikrein related peptidase 3; kallikrein-3; Kallikrein-3 (KLK3); kallikrein-related peptidase 3; Kalp1; KALP15; Kininogenin; KLK2A1; Klk3; KLK-3; Klkb1; P-30 antigen; Pk; Plasma kallikrein; Plasma kallikrein heavy chain; Plasma kallikrein light chain; Plasma prekallikrein; prostate specific antigen; prostate-specific (APS); prostate-specific antigen; PSA; Semenogelase; seminin
Mouse
Size-exclusion chromatography
RUO
354
4°C, do not freeze