Learn More
PSAT1 Rabbit, anti-Human, CoraLite Plus 488, Clone: 1O22, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand:  Proteintech CL488-81479-20UL
 View more versions of this product
Product Code. 18088065
  • 145.00 € / 20µL
Description
The PSAT1 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human PSAT1. This antibody recognizes human, mouse, pig antigen. The PSAT1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
PSAT1
1O22
CoraLite Plus 488
Rabbit
Primary
Human, Mouse, Pig
150 kDa
IgG
Immunofluorescence
1000 μg/mL
EPIP, Phosphoserine aminotransferase, PSA, PSAT, PSAT1
20 μL
29968
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.