missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ PSC Cardiomyocyte Maintenance Medium

Description
Gibco™ Cardiomyocyte Maintenance Medium is a serum-free and xeno-free medium that is capable of maintaining cardiomyocytes that have been differentiated using the PSC Cardiomyocyte Differentiation Kit (Cat. No. A2921201). This medium is included in the kit and sold separately for researchers wishing to maintain differentiated cardiomyocytes in culture for extended periods of time.
- No thawing or mixing required—simply warm and add to differentiated cardiomyocyte culture
- Long-term culture—maintain differentiated cardiomyocytes in culture for >15 days
- Stable phenotype—maintain phenotype throughout long-term culture experiments
Specifications
Specifications
| Cell Type | Cardiomyocytes |
| Product Type | Maintenance Medium |
| Serum Level | Serum-free |
| Quantity | 500 mL |
| Shipping Condition | Room Temperature |
| Culture Type | Mammalian Cell Culture |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?