missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PSD3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15545302
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15545302 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15545302 Supplier Invitrogen™ Supplier No. PA542177

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: SDVAKHLGKN NEFSKLVAEE YLKFFDFTGM TLDQSLRYFF KAFSLVGETQ Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

Guanine nucleotide exchange factor for ARF6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSD3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PSD3
Gene Accession No. Q9NYI0
Gene Alias 4931420C21Rik; ADP-ribosylation factor guanine nucleotide factor 6; AI661273; BC003498; D430018P08; EFA6D; EFA6R; Epididymis tissue protein Li 20mP; exchange factor for ADP-ribosylation factor guanine nucleotide factor 6; Exchange factor for ADP-ribosylation factor guanine nucleotide factor 6 D; Exchange factor for ARF6 D; guanine nucleotide exchange factor for ADP ribosylation factor 6; HCA67; Hepatocellular carcinoma-associated antigen 67; KIAA0942; PH and SEC7 domain-containing protein 3; pleckstrin and Sec7 domain containing 3; pleckstrin homology and SEC7 domain-containing protein 3; Psd3; RGD1559968
Gene Symbols PSD3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human PSD3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23362
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.