Learn More
Thermo Scientific™ Pseudomonas C.F.C. Selective Agar
Description
Selectively isolate Pseudomonas species from food, water, pharmaceutical and environmental samples with Thermo Scientific™ Oxoid™ Pseudomonas C.F.C. Selective Agar. The medium is designed to be selective for Pseudomonas aeruginosa or Pseudomonas spp. generally. It is a modification of King's A Medium1 and contains magnesium chloride and potassium sulphate to enhance the production of pigments. The medium also allows detection of Burkholderia cepacia (formerly Pseudomonas cepacia), an important opportunistic pathogen in water systems 3.
Use Pseudomonas C.F.C. Selective Agar for the isolation of Pseudomonas spp.
- Select for Pseudomonas aeruginosa or Pseudomonas spp.
- Allows detection of Burkholderia cepacia within water systems used for pharmaceutical and cosmetic preparation.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Color | Straw |
| Content And Storage | 2°C to 10°C |
| Description | Pseudomonas C.F.C. Selective Agar |
| Format | 90mm Monoplate |
| Manufacturing Origin | United Kingdom |
| Product Type | Agar Plate |
| Fill Weight | 19.5g±1.0.g |
| Packaging Type | 10 x 90mm plates, film wrapped |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.