missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This antibody has been tested in indirect ELISA.
Human pregnancy-specific glycoproteins (PSG), are a group of closely related secreted glycoproteins which are highly expressed in placental syncytiotrophoblast cells of fetal origin. PSG are commonly expressed in tumors of trophoblast origin (hydatidiform mole, choriocarcinoma). They represent the most abundant fetal protein in the maternal blood at term. The eleven human PSG genes (PSG1-PSG11) form together with the carcinoembryonic antigen (CEA)-related cell adhesion molecule (CEACAM) genes the CEACAM gene family. PSG stimulates secretion of TH2-type cytokines from monocytes. CD9 was shown to represent the monocyte receptor for murine PSG17. PSG are thought to modulate the maternal immune system during pregnancy thus protecting the semiallotypic fetus from rejection.
Specifications
Specifications
| Antigen | PSG1 |
| Applications | ELISA, Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | 5C6G7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | PSG1 |
| Gene Accession No. | P11464 |
| Gene Alias | B1G1; CD66 antigen-like family member F; CD66f; DHFRP2; Fetal liver non-specific cross-reactive antigen 1/2; FLJ90598; FLJ90654; FL-NCA-1/2; PBG1; pregnancy specific beta-1-glycoprotein 1; pregnancy-specific B-1 glycoprotein; pregnancy-specific beta-1 glycoprotein C/D; Pregnancy-specific beta-1-glycoprotein 1; PS-beta-C/D; PS-beta-G-1; PSBG1; PSBG-1; PSG 1; PSG IIA-d; PSG1; PSG-1d; PSG95; PSGGA; PSGIIA; PSGIIA-a; PSGIIA-b; PSGIIA-c; SP 1; SP1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
