missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSG7 Polyclonal Antibody

Description
PSG7 Polyclonal Antibody for Western Blot, IHC (P), Flow
PSG7 is a member of the pregnancy-specific glycoprotein (PSG) gene family. The PSG genes are a subgroup of the carcinoembryonic antigen (CEA) family of immunoglobulin-like genes, and are found in a gene cluster at 19q13.1-q13.2 telomeric to another cluster of CEA-related genes. The PSG genes are expressed by placental trophoblasts and released into the maternal circulation during pregnancy, and are thought to be essential for maintenance of normal pregnancy. The reference genome contains a nonsense mutation that disrupts the coding sequence, suggesting that this gene may be evolving into a pseudogene.
Specifications
Specifications
| Antigen | PSG7 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | PSG7 |
| Gene Accession No. | Q13046 |
| Gene Alias | pregnancy specific beta-1-glycoprotein 7 (gene/pseudogene); pregnancy-specific glycoprotein 7; PS-beta-G-7; PSBG-7; PSG1; PSG7; PSGGA; putative pregnancy-specific beta-1-glycoprotein 7 |
| Gene Symbols | PSG7 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?