Learn More
PSMA/FOLH1/NAALADase I Antibody (CL9091) [DyLight 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-88925V
Product Code. 30301150
  • 737.00 € / 0.10mL
Description
PSMA/FOLH1/NAALADase I Monoclonal antibody specifically detects PSMA/FOLH1/NAALADase I in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PSMA/FOLH1/NAALADase I
Monoclonal
DyLight 405
Cell growth-inhibiting gene 27 protein, cell growth-inhibiting protein 27, EC 3.4.17.21, folate hydrolase (prostate-specific membrane antigen) 1, Folate hydrolase 1, FOLHNAALADase I, Folylpoly-gamma-glutamate carboxypeptidase, GCP2, GCPIImGCP, glutamate carboxylase II, glutamate carboxypeptidase 2, Glutamate carboxypeptidase II, Membrane glutamate carboxypeptidase, NAALAD1N-acetylated-alpha-linked acidic dipeptidase I, NAALAdase, N-acetylated alpha-linked acidic dipeptidase 1, prostate specific membrane antigen variant F, Prostate-specific membrane antigen, PSMAFGCP, PSMPteroylpoly-gamma-glutamate carboxypeptidase
This antibody was developed using a recombinant protein derived from Q04609, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Prostate Cancer
2346
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL9091
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified