Learn More
PSMA4 Monoclonal Antibody (OTI1H10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA504023
Encompass_Preferred
 View more versions of this product
Product Code. 17560602
  • 540.87 € / 100µL
Description
The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the peptidase T1A family, that is a 20S core alpha subunit. Three alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
PSMA4
Monoclonal
0.49 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P21670, P25789, Q9R1P0
Psma4
Full length human recombit protein of human PSMA4 produced in HEK293T cell.
100 μL
Primary
Canine, Human, Mouse, Rat
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI1H10
Unconjugated
PSMA4
HC9, HsT17706, PSC9
Mouse
Affinity Chromatography
RUO
100687556, 26441, 29671, 5685
-20° C, Avoid Freeze/Thaw Cycles