missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMA4 Monoclonal Antibody (OTI5D8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504058S
This item is not returnable.
View return policy
Description
PSMA4 Monoclonal antibody specifically detects PSMA4 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| PSMA4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q15475 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6495 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5D8 | |
| Unconjugated | |
| SIX1 | |
| C9; HC9; HsT17706; LOW QUALITY PROTEIN: proteasome subunit alpha type-4; macropain subunit C9; MGC111191; MGC12467; MGC24813; Multicatalytic endopeptidase complex subunit C9; multicatalytic proteinase subunit L; proteasome (prosome, macropain) subunit, alpha type 4; proteasome (prosome, macropain) subunit, alpha type, 4; proteasome 20S subunit alpha 4; proteasome alpha 4 subunit; proteasome component C9; proteasome subunit alpha 4; proteasome subunit alpha type-4; proteasome subunit alpha type-4-like protein; proteasome subunit HC9; proteasome subunit L; PSC9; PSMA 4; PSMA4; QtsA-10816; unnamed protein product; wu:fe05d10; zgc:56176 | |
| Human recombinant protein fragment corresponding to amino acids 1-284 of human SIX1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction