missing translation for 'onlineSavingsMsg'
Learn More

PSMB8 Rabbit, anti-Human, CoraLite 594, Polyclonal, Proteintech

Product Code. p-200049911 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18008955 100 μL 100µL
18076405 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18008955 Supplier Proteintech Supplier No. CL59414859100UL
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The PSMB8 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PSMB8. This antibody recognizes human, mouse antigen. The PSMB8 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.

Specifications

Antigen PSMB8
Applications Immunofluorescence
Concentration 1000 μg/mL
Conjugate CoraLite 594
Gene Symbols beta5i, D6S216, D6S216E, LMP7, Low molecular mass protein 7, Macropain subunit C13, Proteasome component C13, Proteasome subunit beta 5i, Proteasome subunit beta type 8, PSMB5i, PSMB8, RING10, Y2
Host Species Rabbit
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 5696
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Antibody Molecular Weight 150 kDa
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.