missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMD12 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
188.00 € - 470.00 €
Specifications
| Antigen | PSMD12 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18676402
|
Novus Biologicals
NBP2-94102-0.02ml |
0.02 mL |
188.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18629731
|
Novus Biologicals
NBP2-94102-0.1ml |
0.1 mL |
470.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PSMD12 Polyclonal antibody specifically detects PSMD12 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| PSMD12 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS (pH 7.3), 50% glycerol | |
| 5718 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 26S proteasome non-ATPase regulatory subunit 12, 26S proteasome regulatory subunit p55, 26S proteasome regulatory subunit RPN5, MGC75406, p55, proteasome (prosome, macropain) 26S subunit, non-ATPase, 12, Rpn5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 197-456 of human PSMD12 (NP_002807.1). KDYIRTQIISKKINTKFFQEENTEKLKLKYYNLMIQLDQHEGSYLSICKHYRAIYDTPCIQAESEKWQQALKSVVLYVILAPFDNEQSDLVHRISGDKKLEEIPKYKDLLKLFTTMELMRWSTLVEDYGMELRKGSLESPATDVFGSTEEGEKRWKDLKNRVVEHNIRIMAKYYTRITMKRMAQLLDLSVDESEAFLSNLVVNKTIFAKVDRLAGIINFQRPKDPNNLLNDWSQKLNSLMSLVNKTTHLIAKEEMIHNLQ | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title