missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSMD4 Recombinant Rabbit Monoclonal Antibody (24GB4205)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557197
This item is not returnable.
View return policy
Description
PSMD4 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes a subunit of the elongation factor-1 complex, which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. This subunit functions as guanine nucleotide exchange factor. It is reported that this subunit interacts with HIV-1 Tat, and thus it represses the translation of host-cell, but not HIV-1, mRNAs. Several alternatively spliced transcript variants encoding multiple isoforms have been found for this gene.
Specifications
| PSMD4 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Psmd4 | |
| 26S proteasome non-ATPase regulatory subunit 4; 26S proteasome regulatory subunit RPN10; 26S proteasome regulatory subunit S5A; AF; Af1; AF-1; angiocidin; anti secretory factor 1; antisecretory factor; antisecretory factor 1; ASF; Mcb1; multiubiquitin chain-binding protein; multiubiquitin-chain-binding protein; proteasome (prosome, macropain) 26S subunit, non-ATPase, 4; proteasome 26S subunit, non-ATPase 4; Psmd4; pUB-R5; RP11-126K1.1; Rpn10; RPN10 homolog; S5A; S5a/antisecretory factor protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 19185, 5710, 83499 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB4205 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O35226, P55036 | |
| Psmd4 | |
| A synthesized peptide derived from human PSMD4 (aa 310-377). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction