missing translation for 'onlineSavingsMsg'
Learn More

Psoralen, 97%, Thermo Scientific Chemicals

Product Code. 15442889
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15442889 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15442889 Supplier Thermo Scientific Alfa Aesar Supplier No. J60042.MC
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Intercalates into DNA; photochemical reagent for the investigation of nucleic acid structure

Intercalates into DNA; photochemical reagent for the investigation of nucleic acid structure

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66-97-7
Molecular Formula C11H6O3
Molecular Weight (g/mol) 186.17
MDL Number MFCD00010520
InChI Key ZCCUUQDIBDJBTK-UHFFFAOYSA-N
Synonym psoralen, ficusin, 7h-furo 3,2-g chromen-7-one, furocoumarin, psoralene, 7h-furo 3,2-g 1 benzopyran-7-one, psorline-p, 6,7-furanocoumarin, furo 3,2-g chromen-7-one, furo 3,2-g coumarin
PubChem CID 6199
ChEBI CHEBI:27616
IUPAC Name 7H-furo[3,2-g]chromen-7-one
SMILES O=C1OC2=CC3=C(C=CO3)C=C2C=C1

Specifications

Melting Point 163°C to 164°C
Color White
Quantity 100 mg
Beilstein 152784
Merck Index 14,7928
Formula Weight 186.16
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Psoralen
compliance-icons
Product Identifier
  • Psoralen
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 7H-Furo[3,2-g][1]benzopyran-7-one

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.