accessibility menu, dialog, popup

UserName

OOO

CAS RN 66-97-7

IUPAC Name: 7H-furo[3,2-g]chromen-7-one
Synonyms: psoralen psoralen (product) furo[2',3':7,6]coumarin furo(4',5',6,7)coumarin psoralene furo(2',3',7,6)coumarin furo[4',5':6,7]coumarin psorline-P psoralen (substance) ficusin
Molecular Weight (g/mol): 186.17
Molecular Formula: C11H6O3
InChi Key: ZCCUUQDIBDJBTK-UHFFFAOYSA-N
SMILES: O=C1OC2=CC3=C(C=CO3)C=C2C=C1
Molecular Weight (g/mol) 
  • (7)
  • (1)
Percent Purity 
  • (3)
  • (1)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)
Color 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results