missing translation for 'onlineSavingsMsg'
Learn More

Psoralen, MedChemExpress

Product Code. 16448848
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16448848 10 mg 10mg
16464737 50 mg 50mg
16448966 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16448848 Supplier Medchem Express Supplier No. HYN005310mg
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Psoralen (Ficusin) is a coumarin isolated from the seeds of Fructus Psoraleae. Psoralen exhibits a wide range of biological properties, including anti-cancer, antioxidant, antidepressant, anticancer, antibacterial, and antiviral, et al.

Psoralen (Ficusin) is a coumarin isolated from the seeds of Fructus Psoraleae. Psoralen exhibits a wide range of biological properties, including anti-cancer, antioxidant, antidepressant, anticancer, antibacterial, and antiviral, et al.

Specifications

Chemical Name or Material Psoralen
Physical Form Solid
CAS 66-97-7
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C11H6O3
For Use With (Application) Cancer-programmed cell death
Synonym Ficusin
Solubility Information DMSO : 100 mg/mL (537.17 mM; Need ultrasonic) ∣H2O : 1 mg/mL (5.37 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C1C=CC2=CC3=C(OC=C3)C=C2O1
Molecular Weight (g/mol) 186.16
Formula Weight 186.16
Percent Purity 99.67%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.